Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000399583.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000457174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000457174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000457174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000469399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000469399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000469399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000472999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000473130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
ENST00000484211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013508.hg.1 |
NM_018059.4 |
Refseq |