Assay Details
Gene Symbol: LOC107987373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724896.2 | XP_006724959.1 | 1 - 2 | 111 | 128 | |
XM_017030111.1 | XP_016885600.1 | 1 - 2 | 111 | 128 |
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021134.3 | NP_066957.3 | 1 - 2 | 111 | 128 | |
XM_006718271.2 | XP_006718334.1 | 1 - 2 | 111 | 128 | ||
XM_011520273.1 | XP_011518575.1 | 1 - 2 | 111 | 128 | ||
XM_011520275.2 | XP_011518577.1 | 1 - 2 | 111 | 128 | ||
XM_011520276.2 | XP_011518578.1 | 1 - 2 | 111 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000055.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_021134.3,XM_006718271.2,XM_006724896.2,XM_011520273.1,XM_011520275.2,XM_011520276.2,XM_017030111.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | ENST00000381514.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | ENST00000381519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | ENST00000397294.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | ENST00000397297.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | ENST00000397298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001098.hg.1 | TC11000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000381514.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000381519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000397294.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000397297.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000397298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001102.hg.1 | TC11000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11001096.hg.1 | ENST00000381519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001096.hg.1 | ENST00000397297.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001096.hg.1 | ENST00000397298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001096.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001096.hg.1 | TC11000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | ENST00000381514.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | ENST00000381519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | ENST00000397297.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | ENST00000397298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001097.hg.1 | TC11000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11001091.hg.1 | ENST00000381519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001091.hg.1 | ENST00000397298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11001091.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11001091.hg.1 | TC11000055.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100145756.hg.1 | TC1100006547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100145743.hg.1 | TC1100006547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
