Assay Details
| Gene Symbol: LRIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312686.1 | NP_001299615.1 | 1 - 2 | 473 | 117 | |
| NM_014813.2 | NP_055628.1 | 1 - 2 | 473 | 117 | ||
| XM_005271369.2 | XP_005271426.1 | 1 - 2 | 473 | 117 | ||
| GenBank mRNA | AB018349.1 | - | 1 - 2 | 434 | 117 | |
| AK291031.1 | - | 1 - 2 | 465 | 117 | ||
| AL161998.1 | - | 1 - 2 | 432 | 117 | ||
| BC117368.1 | - | 1 - 2 | 297 | 40125919 | 117 | |
| BC117370.1 | - | 1 - 2 | 297 | 40125921 | 117 | |
| DA506094.1 | - | 1 - 2 | 473 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000998.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001312686.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01015882.hg.1 | TC01000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015882.hg.1 | ENST00000361127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015882.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015883.hg.1 | TC01000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015883.hg.1 | ENST00000361127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015883.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015881.hg.1 | TC01000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015881.hg.1 | ENST00000361127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015881.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168181.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168181.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168181.hg.1 | TC0100009468.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100168184.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168184.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168184.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168184.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168184.hg.1 | TC0100009468.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100168186.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168186.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168186.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168186.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168186.hg.1 | TC0100009468.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168182.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168182.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168182.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168182.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168182.hg.1 | TC0100009468.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100168183.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168183.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168183.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168183.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168183.hg.1 | TC0100009468.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 103013 | - | - |

LRIG2
Genomic Map