Assay Details
| Gene Symbol: PHTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323042.1 | NP_001309971.1 | 1 - 2 | 120 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323042.1 transcripts.
Set Membership: |
|

PHTF1
Genomic Map