Assay Details
| Gene Symbol: CHP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK307060.1 | - | 1 - 2 | 184 | 63 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009176.hg.1 | ENST00000427102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009176.hg.1 | ENST00000427102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000334660.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000392151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000558351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000560965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | ENST00000561280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006994.hg.1 | NM_007236.4 | Refseq |

CHP1
Genomic Map