Assay Details
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058004.3 | NP_477352.3 | 24 - 25 | 2949 | 63 | |
| XM_005261634.1 | XP_005261691.1 | 24 - 25 | 2949 | 63 | ||
| XM_005261635.1 | XP_005261692.1 | 24 - 25 | 2949 | 63 | ||
| XM_011530226.1 | XP_011528528.1 | 24 - 25 | 2949 | 63 | ||
| XM_011530228.2 | XP_011528530.1 | 24 - 25 | 2949 | 63 | ||
| XM_017028829.1 | XP_016884318.1 | 23 - 24 | 2883 | 63 | ||
| XM_017028830.1 | XP_016884319.1 | 10 - 11 | 1105 | 63 | ||
| GenBank mRNA | AB210002.1 | - | 24 - 25 | 2919 | 63 | |
| AF012872.1 | - | 23 - 24 | 2688 | 63 | ||
| AK302424.1 | - | 24 - 25 | 2877 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001433.hg.1
Set Membership: |
|

PI4KA
Genomic Map