Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008515.hg.1 |
ENST00000323776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008515.hg.1 |
ENST00000436259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008515.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008515.hg.1 |
TC17000669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008514.hg.1 |
ENST00000323776.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008514.hg.1 |
ENST00000436259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008514.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008514.hg.1 |
TC17000669.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000323776.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000436259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000437498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000488432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000323776.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000436259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000437498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
ENST00000488432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161280.hg.1 |
TC1700008259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000323776.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000436259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000437498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000488432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000323776.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000436259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000437498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
ENST00000488432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
NM_032133.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161279.hg.1 |
TC1700008259.hg.1 |
Clariom D TC |