Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
ENST00000335894.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
ENST00000358767.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
ENST00000360277.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
ENST00000546225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
ENST00000558759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
NM_001031715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005142.hg.1 |
TC15000623.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000335894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000358767.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000514049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000546225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000558759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001031715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001284347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001284348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000335894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000358767.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000514049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000546225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
ENST00000558759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001031715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001284347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
NM_001284348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154227.hg.1 |
TC1500007640.hg.1 |
Clariom D TC |