Assay Details
| Gene Symbol: IQCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031715.2 | NP_001026885.1 | 5 - 6 | 605 | 88 | |
| XM_011521907.2 | XP_011520209.1 | 6 - 7 | 746 | 88 | ||
| XM_011521908.2 | XP_011520210.1 | 5 - 6 | 605 | 88 | ||
| XM_011521911.2 | XP_011520213.1 | 4 - 5 | 533 | 88 | ||
| XM_011521912.2 | XP_011520214.1 | 6 - 7 | 746 | 88 | ||
| XM_011521913.2 | XP_011520215.1 | 6 - 7 | 747 | 88 | ||
| XM_011521914.2 | XP_011520216.1 | 3 - 4 | 265 | 88 | ||
| XM_011521915.2 | XP_011520217.1 | 6 - 7 | 746 | 88 | ||
| XM_011521922.2 | XP_011520224.1 | 6 - 7 | 749 | 88 | ||
| XM_017022491.1 | XP_016877980.1 | 6 - 7 | 746 | 88 | ||
| XM_017022492.1 | XP_016877981.1 | 6 - 7 | 746 | 88 | ||
| XM_017022493.1 | XP_016877982.1 | 6 - 7 | 746 | 88 | ||
| XM_017022494.1 | XP_016877983.1 | 6 - 7 | 746 | 88 | ||
| XM_017022495.1 | XP_016877984.1 | 6 - 7 | 746 | 88 | ||
| XM_017022496.1 | XP_016877985.1 | 6 - 7 | 746 | 88 | ||
| XM_017022497.1 | XP_016877986.1 | 6 - 7 | 746 | 88 | ||
| XM_017022498.1 | XP_016877987.1 | 6 - 7 | 746 | 88 | ||
| XM_017022499.1 | XP_016877988.1 | 6 - 7 | 746 | 88 | ||
| GenBank mRNA | AK292478.1 | - | 5 - 6 | 557 | 88 | |
| AY014282.2 | - | 5 - 6 | 576 | 88 | ||
| BC048964.1 | - | 5 - 6 | 555 | 5297681 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

IQCH
Genomic Map