Assay Details
| Gene Symbol: IDH3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004135.3 | NP_004126.1 | 2 - 4 | 325 | 65 | |
| NM_174869.2 | NP_777358.1 | 2 - 4 | 325 | 65 | ||
| GenBank mRNA | BC000933.2 | - | 2 - 4 | 171 | 3447003 | 65 |
| BC001902.1 | - | 2 - 4 | 166 | 3543839 | 65 | |
| BI831096.1 | - | 2 - 4 | 325 | 65 | ||
| U40272.1 | - | 2 - 4 | 320 | 65 | ||
| Z68907.1 | - | 2 - 4 | 207 | 65 | ||
More Information
Set Membership: |
|

IDH3G
Genomic Map