Assay Details
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005253488.2 | XP_005253545.1 | 3 - 4 | 1455 | 70 | |
XM_017019988.1 | XP_016875477.1 | 3 - 4 | 858 | 70 | ||
GenBank mRNA | AY038927.1 | - | 2 - 3 | 394 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
