Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102371.1 | NP_001095841.1 | 6 - 7 | 1474 | 84 | |
| NM_024955.5 | NP_079231.4 | 6 - 7 | 1513 | 84 | ||
| XM_017028941.1 | XP_016884430.1 | 5 - 6 | 1146 | 84 | ||
| XM_017028942.1 | XP_016884431.1 | 5 - 6 | 1243 | 84 | ||
| GenBank mRNA | AK025714.1 | - | 6 - 7 | 1413 | 84 | |
| AK026975.1 | - | 3 - 4 | 594 | 84 | ||
| AK225347.1 | - | 6 - 7 | 1413 | 84 | ||
| AK315555.1 | - | 6 - 7 | 1416 | 84 | ||
| BC015726.1 | - | 6 - 7 | 1397 | 4866761 | 84 | |
| BC027716.2 | - | 6 - 7 | 1492 | 4127473 | 84 | |
| BC040351.1 | - | 6 - 7 | 1436 | 3873448 | 84 | |
| CR456444.1 | - | 5 - 6 | 1488 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000710.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | ENST00000216187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | ENST00000366463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | ENST00000397224.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012498.hg.1 | TC22000710.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | TC2200008610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162027.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |

FOXRED2
Genomic Map