| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
TC07000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000265437.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000323984.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393446.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393447.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000393451.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000422922.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000432298.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000465133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
ENST00000487459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
NM_018412.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011560.hg.1 |
NM_021908.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000265437.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000323984.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393446.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393447.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393451.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000422922.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000432298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000438863.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000446490.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000465133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000487459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000489293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000490039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
NM_018412.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
NM_021908.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000265437.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000323984.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393446.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393447.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000393451.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000422922.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000432298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000438863.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000446490.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000465133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000487459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000489293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
ENST00000490039.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
NM_018412.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
NM_021908.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162945.hg.1 |
TC0700008882.hg.1 |
Clariom D TC |