Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
TC07001722.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
ENST00000343529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
ENST00000424685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
ENST00000424828.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
ENST00000428762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
ENST00000448171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
NM_005045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026720.hg.1 |
NM_173054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000343529.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000424685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000428762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
NM_005045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
NM_173054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000343529.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000424685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
ENST00000428762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
NM_005045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
NM_173054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187348.hg.1 |
TC0700012126.hg.1 |
Clariom D TC |