Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000401.hg.1 |
TC19002616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000401.hg.1 |
ENST00000215368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000401.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900198796.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900198796.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900198796.hg.1 |
TC1900011640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900198796.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900198796.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
ENST00000215368.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011640.hg.1 |
NM_001405.3 |
Refseq |