Assay Details
| Gene Symbol: TTPAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024331.4 | NP_077307.2 | 1 - 2 | 105 | 73 | |
| XM_017028063.1 | XP_016883552.1 | 1 - 2 | 96 | 73 | ||
| GenBank mRNA | BC003071.2 | - | 1 - 2 | 84 | 2989837 | 73 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000333.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC003071.2,NM_024331.4,XM_017028063.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | ENST00000262605.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | ENST00000372904.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004711.hg.1 | TC20000333.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | ENST00000262605.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | ENST00000372904.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004715.hg.1 | TC20000333.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | TC2000007456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151734.hg.1 | NM_024331.4 | Refseq |

TTPAL
Genomic Map