Assay Details
| Gene Symbol: TTPAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024331.4 | NP_077307.2 | 2 - 3 | 132 | 65 | |
| XM_017028063.1 | XP_016883552.1 | 2 - 3 | 123 | 65 | ||
| GenBank mRNA | BC003071.2 | - | 2 - 3 | 111 | 2989837 | 65 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003071.2, NM_024331.4, XM_017028063.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | TC2000007456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151739.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000262605.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000372906.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000456317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | ENST00000461134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001039199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_001261839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007456.hg.1 | NM_024331.4 | Refseq |

TTPAL
Genomic Map