Assay Details
| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016067.3 | NP_057151.1 | 3 - 4 | 377 | 144 | |
| GenBank mRNA | AF151892.1 | - | 3 - 4 | 290 | 144 | |
| AK026620.1 | - | 3 - 4 | 344 | 144 | ||
| BC005186.1 | - | 3 - 4 | 268 | 3839381 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MRPS18C
Genomic Map