Assay Details
Gene Symbol: ZFP36L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK024202.1 | - | 1 | 437 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK024202.1 transcripts.
This assay is in the 5'UTR region of the AK024202.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 230520 | - | - |
