Assay Details
| Gene Symbol: REPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128617.2 | NP_001122089.1 | 9 - 10 | 1836 | 64 | |
| NM_001286612.1 | NP_001273541.1 | 9 - 10 | 1836 | 64 | ||
| XM_005267179.3 | XP_005267236.1 | 9 - 10 | 1857 | 64 | ||
| XM_017011389.1 | XP_016866878.1 | 9 - 10 | 1223 | 64 | ||
| GenBank mRNA | AL832307.1 | - | 8 - 9 | 1082 | 64 | |
| BC021211.2 | - | 9 - 10 | 1456 | 4136619 | 64 | |
More Information
Set Membership: |
|

REPS1
Genomic Map