Assay Details
Gene Symbol: KHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251874.1 | NP_001238803.1 | 2 - 3 | 709 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | TC06001869.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | ENST00000257765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | ENST00000370384.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | ENST00000370385.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | ENST00000423730.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | NM_001251874.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06022653.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014311.hg.1 | ENST00000511053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000257765.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000433730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | ENST00000610435.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014313.hg.1 | NM_030568.4 | Refseq |
