Assay Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318737.1 | NP_001305666.1 | 1 - 2 | 82 | 116 | |
NM_024725.3 | NP_079001.2 | 1 - 2 | 82 | 116 | ||
XM_006718911.3 | XP_006718974.1 | 1 - 2 | 103 | 116 | ||
XM_011542987.2 | XP_011541289.1 | 1 - 2 | 94 | 116 | ||
XM_011542988.2 | XP_011541290.1 | 1 - 2 | 94 | 116 | ||
GenBank mRNA | AK056790.1 | - | 1 - 2 | 75 | 116 | |
BP219605.1 | - | 1 - 2 | 107 | 116 | ||
DA801410.1 | - | 1 - 2 | 82 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002212.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK056790.1,NM_001318737.1,NM_024725.3,XM_006718911.3,XM_011542987.2,XM_011542988.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11026517.hg.1 | ENST00000278520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026517.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026517.hg.1 | ENST00000542662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026517.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026517.hg.1 | TC11002212.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11026521.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026521.hg.1 | ENST00000542662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026521.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026521.hg.1 | TC11002212.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192813.hg.1 | TC1100012052.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100192816.hg.1 | TC1100012052.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 174537 | - | - |
