| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005641.hg.1 |
TC01000362.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005641.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005641.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005639.hg.1 |
TC01000362.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005639.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005639.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152593.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152593.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152593.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152593.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152593.hg.1 |
TC0100007512.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007512.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152595.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152595.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152595.hg.1 |
ENST00000374024.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152595.hg.1 |
NM_005281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152595.hg.1 |
TC0100007512.hg.1 |
Clariom D TC |