Assay Details
| Gene Symbol: LOC105377443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U79300.1 | - | 1 | 802 | 187 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin 18 | PCDH18 | PCDH68L | -69.5kb | Anti-sense | Chr. 4 - 137518918 - 137532530 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 616 | LINC00616 | 424.0kb | Anti-sense | Chr. 4 - 138027422 - 138130685 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105377443
Genomic Map