Assay Details
Gene Symbol: ADARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018702.3 | NP_061172.1 | 7 - 8 | 2006 | 96 | |
GenBank mRNA | AF034837.1 | - | 7 - 8 | 2014 | 96 | |
BC137477.1 | - | 7 - 8 | 1827 | 9021477 | 96 | |
BC140852.1 | - | 7 - 8 | 1827 | 9021720 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000969.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10012168.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012168.hg.1 | ENST00000381310.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012168.hg.1 | ENST00000381312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012168.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012168.hg.1 | TC10000969.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381305.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000381312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | ENST00000477140.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | NM_018702.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000165194.hg.1 | TC1000009541.hg.1 | Clariom D TC |
