Assay Details
| Gene Symbol: LRIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312686.1 | NP_001299615.1 | 4 - 5 | 734 | 265 | |
| NM_014813.2 | NP_055628.1 | 3 - 4 | 622 | 153 | ||
| XM_005271369.2 | XP_005271426.1 | 3 - 4 | 622 | 153 | ||
| XM_017002952.1 | XP_016858441.1 | 2 - 3 | 146 | 153 | ||
| GenBank mRNA | AB018349.1 | - | 3 - 4 | 583 | 153 | |
| AK291031.1 | - | 3 - 4 | 614 | 153 | ||
| BC117368.1 | - | 3 - 4 | 446 | 40125919 | 153 | |
| BC117370.1 | - | 3 - 4 | 446 | 40125921 | 153 | |
| CR936669.1 | - | 4 - 5 | 725 | 265 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000998.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001312686.1,XM_017002952.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015886.hg.1 | TC01000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015886.hg.1 | ENST00000361127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015886.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000466161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000470000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | ENST00000492207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009468.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168190.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168190.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168190.hg.1 | ENST00000361127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168190.hg.1 | NM_014813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168190.hg.1 | TC0100009468.hg.1 | Clariom D TC |

LRIG2
Genomic Map