Assay Details
| Gene Symbol: PRRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022716.3 | NP_073207.1 | 3 - 4 | 649 | 82 | |
| XM_006711388.3 | XP_006711451.1 | 4 - 5 | 712 | 82 | ||
| GenBank mRNA | AB451463.1 | - | 3 - 4 | 602 | 82 | |
| BC074993.2 | - | 3 - 4 | 649 | 30915308 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001484.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of mesenchymal cell proliferation
embryonic limb morphogenesis
inner ear morphogenesis
middle ear morphogenesis
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
neuron fate determination
embryonic cranial skeleton morphogenesis
artery morphogenesis
cartilage development
palate development
regulation of neuron projection regeneration
neuronal stem cell population maintenance
RNA polymerase II transcription coactivator activity
transcription coactivator activity
sequence-specific DNA binding
HMG box domain binding
negative regulation of transcription from RNA polymerase II promoter
positive regulation of mesenchymal cell proliferation
embryonic limb morphogenesis
inner ear morphogenesis
middle ear morphogenesis
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
neuron fate determination
embryonic cranial skeleton morphogenesis
artery morphogenesis
cartilage development
palate development
regulation of neuron projection regeneration
neuronal stem cell population maintenance
RNA polymerase II transcription coactivator activity
transcription coactivator activity
sequence-specific DNA binding
HMG box domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | TC01001484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | ENST00000239461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | ENST00000367760.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | ENST00000476867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | ENST00000495280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022994.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000497230.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | ENST00000553786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010571.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000239461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000367760.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000476867.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000485529.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000495280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | ENST00000496573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | NM_006902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | NM_022716.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177794.hg.1 | TC0100010571.hg.1 | Clariom D TC |

PRRX1
Genomic Map