Assay Details
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006943.1 | NP_001006944.1 | 4 - 5 | 1108 | 98 | |
XM_011540969.1 | XP_011539271.1 | 6 - 7 | 1271 | 98 | ||
XM_011540970.1 | XP_011539272.1 | 4 - 5 | 1005 | 98 | ||
XM_011540972.1 | XP_011539274.1 | 5 - 6 | 1172 | 98 | ||
XM_011540973.1 | XP_011539275.1 | 3 - 4 | 907 | 98 | ||
GenBank mRNA | AB040892.1 | - | 2 - 3 | 529 | 98 | |
BC038796.1 | - | 4 - 5 | 1038 | 98 | ||
BC072417.1 | - | 4 - 5 | 1108 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000284.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
substrate-dependent cell migration
cell adhesion
axon guidance
neuron remodeling
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron projection development
regulation of cell adhesion mediated by integrin
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
protein autophosphorylation
ephrin receptor signaling pathway
GPI-linked ephrin receptor activity
ATP binding
substrate-dependent cell migration
cell adhesion
axon guidance
neuron remodeling
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron projection development
regulation of cell adhesion mediated by integrin
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
protein autophosphorylation
ephrin receptor signaling pathway
GPI-linked ephrin receptor activity
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | TC01000284.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | ENST00000166244.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | ENST00000374644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004359.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004360.hg.1 | TC01000284.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004360.hg.1 | ENST00000374644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004360.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004360.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | TC01000284.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | ENST00000166244.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | ENST00000374644.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004358.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150735.hg.1 | TC0100007288.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150737.hg.1 | TC0100007288.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150734.hg.1 | TC0100007288.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100150736.hg.1 | TC0100007288.hg.1 | Clariom D TC |
