Assay Details
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540969.1 | XP_011539271.1 | 9 - 10 | 1886 | 79 | |
XM_011540970.1 | XP_011539272.1 | 7 - 8 | 1620 | 79 | ||
XM_011540972.1 | XP_011539274.1 | 8 - 9 | 1787 | 79 | ||
XM_011540973.1 | XP_011539275.1 | 6 - 7 | 1522 | 79 | ||
XM_011540975.2 | XP_011539277.1 | 4 - 5 | 698 | 79 | ||
GenBank mRNA | AB040892.1 | - | 5 - 6 | 1144 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
substrate-dependent cell migration
cell adhesion
axon guidance
neuron remodeling
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron projection development
regulation of cell adhesion mediated by integrin
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
protein autophosphorylation
ephrin receptor signaling pathway
GPI-linked ephrin receptor activity
ATP binding
substrate-dependent cell migration
cell adhesion
axon guidance
neuron remodeling
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron projection development
regulation of cell adhesion mediated by integrin
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
protein autophosphorylation
ephrin receptor signaling pathway
GPI-linked ephrin receptor activity
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000166244.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000374644.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | ENST00000538803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_001006943.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007288.hg.1 | NM_020526.4 | Refseq |
