Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015289.hg.1 |
TC01000957.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015289.hg.1 |
ENST00000369795.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015289.hg.1 |
ENST00000369796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015289.hg.1 |
ENST00000485775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015289.hg.1 |
NM_033088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000369795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000369796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000485775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NM_001270768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NM_033088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NR_073071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000369795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000369796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
ENST00000485775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NM_001270768.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NM_033088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
NR_073071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167224.hg.1 |
TC0100009366.hg.1 |
Clariom D TC |