Assay Details
| Gene Symbol: ARR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012096.1 | - | 15 - 16 | 1163 | 4645867 | 77 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151608.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151608.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151608.hg.1 | TC0X00007535.hg.1 | Clariom D TC |

ARR3
Genomic Map