Assay Details
Gene Symbol: PCSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000439.4 | NP_000430.3 | 11 - 12 | 1830 | 127 | |
NM_001177875.1 | NP_001171346.1 | 11 - 12 | 1488 | 127 | ||
GenBank mRNA | AB208874.1 | - | 11 - 12 | 1506 | 127 | |
AK303888.1 | - | 11 - 12 | 1488 | 127 | ||
AK308575.1 | - | 5 - 6 | 736 | 127 | ||
BC130295.1 | - | 11 - 12 | 1712 | 40146326 | 127 | |
BC136486.1 | - | 11 - 12 | 1712 | 9020475 | 127 | |
M90753.1 | - | 11 - 12 | 1800 | 127 | ||
X64810.1 | - | 11 - 12 | 1780 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001614.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | TC05001614.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | ENST00000311106.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | ENST00000508626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05022026.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000311106.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000508626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | ENST00000513085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | NM_000439.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | NM_001177875.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179791.hg.1 | TC0500011513.hg.1 | Clariom D TC |
