Assay Details
Gene Symbol: GCGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000160.4 | NP_000151.1 | 5 - 6 | 686 | 64 | |
XM_006722277.1 | XP_006722340.1 | 5 - 6 | 715 | 64 | ||
XM_011523539.1 | XP_011521841.1 | 5 - 6 | 813 | 64 | ||
XM_017024446.1 | XP_016879935.1 | 5 - 6 | 734 | 64 | ||
XM_017024447.1 | XP_016879936.1 | 4 - 5 | 523 | 64 | ||
GenBank mRNA | AK314247.1 | - | 5 - 6 | 669 | 64 | |
BC104854.1 | - | 4 - 5 | 436 | 8143857 | 64 | |
BC112041.1 | - | 4 - 5 | 435 | 8327509 | 64 | |
L20316.1 | - | 5 - 6 | 668 | 64 | ||
U03469.1 | - | 4 - 5 | 452 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000937.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017024447.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
exocytosis
cell surface receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
response to nutrient
regulation of blood pressure
hormone-mediated signaling pathway
glucose homeostasis
response to starvation
positive regulation of GTPase activity
regulation of glycogen metabolic process
cellular response to glucagon stimulus
glucagon receptor activity
guanyl-nucleotide exchange factor activity
peptide hormone binding
generation of precursor metabolites and energy
exocytosis
cell surface receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
response to nutrient
regulation of blood pressure
hormone-mediated signaling pathway
glucose homeostasis
response to starvation
positive regulation of GTPase activity
regulation of glycogen metabolic process
cellular response to glucagon stimulus
glucagon receptor activity
guanyl-nucleotide exchange factor activity
peptide hormone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17012353.hg.1 | ENST00000400723.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17012353.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17012353.hg.1 | TC17000937.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700169771.hg.1 | TC1700009194.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
