Assay Details
| Gene Symbol: GCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000160.4 | NP_000151.1 | 6 - 7 | 800 | 116 | |
| XM_006722277.1 | XP_006722340.1 | 6 - 7 | 829 | 116 | ||
| XM_011523539.1 | XP_011521841.1 | 6 - 7 | 927 | 116 | ||
| XM_017024446.1 | XP_016879935.1 | 6 - 7 | 848 | 116 | ||
| XM_017024447.1 | XP_016879936.1 | 5 - 6 | 637 | 116 | ||
| GenBank mRNA | AK314247.1 | - | 6 - 7 | 783 | 116 | |
| BC104854.1 | - | 5 - 6 | 550 | 8143857 | 116 | |
| BC112041.1 | - | 5 - 6 | 549 | 8327509 | 116 | |
| L20316.1 | - | 6 - 7 | 782 | 116 | ||
| U03469.1 | - | 5 - 6 | 566 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000937.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
exocytosis
cell surface receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
response to nutrient
regulation of blood pressure
hormone-mediated signaling pathway
glucose homeostasis
response to starvation
positive regulation of GTPase activity
regulation of glycogen metabolic process
cellular response to glucagon stimulus
glucagon receptor activity
guanyl-nucleotide exchange factor activity
peptide hormone binding
generation of precursor metabolites and energy
exocytosis
cell surface receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
response to nutrient
regulation of blood pressure
hormone-mediated signaling pathway
glucose homeostasis
response to starvation
positive regulation of GTPase activity
regulation of glycogen metabolic process
cellular response to glucagon stimulus
glucagon receptor activity
guanyl-nucleotide exchange factor activity
peptide hormone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17012354.hg.1 | ENST00000400723.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012354.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012354.hg.1 | TC17000937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000571425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000572185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000573428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | ENST00000574283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009194.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | ENST00000400723.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | ENST00000570996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | NM_000160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169773.hg.1 | TC1700009194.hg.1 | Clariom D TC |

GCGR
Genomic Map