Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297716.1 | NP_001284645.1 | 11 - 12 | 2037 | 74 | |
| NM_014979.3 | NP_055794.3 | 11 - 12 | 2037 | 74 | ||
| XM_011543281.2 | XP_011541583.1 | 11 - 12 | 2404 | 74 | ||
| XM_011543282.2 | XP_011541584.2 | 11 - 12 | 2964 | 74 | ||
| XM_017009244.1 | XP_016864733.1 | 7 - 8 | 955 | 74 | ||
| GenBank mRNA | AB028977.1 | - | 9 - 10 | 1098 | 74 | |
| AK094917.1 | - | 11 - 12 | 2038 | 74 | ||
| BC100824.1 | - | 11 - 12 | 1973 | 40001296 | 74 | |
| BC100825.1 | - | 11 - 12 | 1971 | 40001297 | 74 | |
| BC100826.1 | - | 11 - 12 | 1971 | 40001302 | 74 | |
| BC100827.1 | - | 11 - 12 | 1971 | 40001303 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000366.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000581282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005121.hg.1 | TC05000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005121.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005121.hg.1 | ENST00000502798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005121.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153485.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153485.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153485.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153485.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153485.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000581282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153484.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153484.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153484.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153484.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153484.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153479.hg.1 | TC0500007820.hg.1 | Clariom D TC |

SV2C
Genomic Map