Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297716.1 | NP_001284645.1 | 2 - 3 | 776 | 66 | |
| NM_014979.3 | NP_055794.3 | 2 - 3 | 776 | 66 | ||
| XM_011543281.2 | XP_011541583.1 | 2 - 3 | 1143 | 66 | ||
| XM_011543282.2 | XP_011541584.2 | 2 - 3 | 1703 | 66 | ||
| GenBank mRNA | AK094917.1 | - | 2 - 3 | 777 | 66 | |
| BC100824.1 | - | 2 - 3 | 712 | 40001296 | 66 | |
| BC100825.1 | - | 2 - 3 | 710 | 40001297 | 66 | |
| BC100826.1 | - | 2 - 3 | 710 | 40001302 | 66 | |
| BC100827.1 | - | 2 - 3 | 710 | 40001303 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000366.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05005104.hg.1 | TC05000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005104.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005104.hg.1 | ENST00000502798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005104.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005105.hg.1 | TC05000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005105.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005105.hg.1 | ENST00000502798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005105.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005107.hg.1 | TC05000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005107.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005107.hg.1 | ENST00000502798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005107.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011182.hg.1 | ENST00000503652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011182.hg.1 | ENST00000503652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | ENST00000581282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005106.hg.1 | TC05000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005106.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005106.hg.1 | ENST00000502798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005106.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011182.hg.1 | ENST00000503652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011182.hg.1 | ENST00000503652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153465.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153466.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153468.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000506257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | ENST00000581282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007820.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153463.hg.1 | TC0500007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | ENST00000322285.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | ENST00000502798.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | NM_001297716.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | NM_014979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153467.hg.1 | TC0500007820.hg.1 | Clariom D TC |

SV2C
Genomic Map