Assay Details
Gene Symbol: CYP20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177538.2 | NP_803882.1 | 6 - 7 | 1302 | 123 | |
XM_005246698.3 | XP_005246755.1 | 5 - 6 | 766 | 123 | ||
XM_005246699.3 | XP_005246756.1 | 4 - 5 | 693 | 123 | ||
XM_006712644.3 | XP_006712707.1 | 6 - 7 | 754 | 123 | ||
GenBank mRNA | AF289579.1 | - | 2 - 3 | 175 | 123 | |
AK297082.1 | - | 6 - 7 | 785 | 123 | ||
AK307931.1 | - | 6 - 7 | 713 | 123 | ||
AL832998.1 | - | 5 - 6 | 549 | 123 | ||
AY359068.1 | - | 6 - 7 | 722 | 123 | ||
BC020616.1 | - | 6 - 7 | 721 | 4689065 | 123 | |
BC033752.1 | - | 6 - 7 | 734 | 5211537 | 123 | |
BC093022.1 | - | 4 - 5 | 544 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001196.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF289579.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
