Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002662.hg.1 |
TC04000174.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002662.hg.1 |
ENST00000264864.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002662.hg.1 |
ENST00000512921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002662.hg.1 |
ENST00000537420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002662.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400187983.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400187983.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400187983.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400187983.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400187983.hg.1 |
TC0400012780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
ENST00000264864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012780.hg.1 |
NM_018323.3 |
Refseq |