Assay Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173546.2 | NP_775817.1 | 1 - 2 | 102 | 77 | |
XM_005264938.2 | XP_005264995.1 | 1 - 2 | 567 | 77 | ||
XM_005264940.4 | XP_005264997.1 | 1 - 2 | 567 | 77 | ||
XM_006713015.2 | XP_006713078.1 | 1 - 2 | 567 | 77 | ||
XM_006713016.2 | XP_006713079.1 | 1 - 2 | 567 | 77 | ||
GenBank mRNA | AK294160.1 | - | 1 - 2 | 89 | 77 | |
AY129011.1 | - | 1 - 2 | 59 | 77 | ||
BC039083.1 | - | 1 - 2 | 52 | 5167173 | 77 | |
T08504.1 | - | 1 - 2 | 102 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000288.hg
Additional Information:
This assay is in the 5'UTR region of the AK294160.1,AY129011.1,BC039083.1,NM_173546.2,XM_005264938.2,XM_005264940.4,XM_006713015.2,XM_006713016.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03005912.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005912.hg.1 | ENST00000332780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005912.hg.1 | ENST00000538729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005912.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005913.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005913.hg.1 | ENST00000332780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005913.hg.1 | ENST00000538729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005913.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005914.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005914.hg.1 | ENST00000332780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005914.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005914.hg.1 | ENST00000538729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005914.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005915.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005915.hg.1 | ENST00000332780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005915.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005915.hg.1 | ENST00000538729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005915.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005909.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005909.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152312.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152312.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152312.hg.1 | TC0300007367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152316.hg.1 | TC0300007367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152317.hg.1 | TC0300007367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152319.hg.1 | TC0300007367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152315.hg.1 | TC0300007367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152318.hg.1 | TC0300007367.hg.1 | Clariom D TC |
