Assay Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173546.2 | NP_775817.1 | 2 - 3 | 609 | 88 | |
XM_005264938.2 | XP_005264995.1 | 2 - 3 | 1074 | 88 | ||
GenBank mRNA | AY129011.1 | - | 2 - 3 | 566 | 88 | |
BC039083.1 | - | 2 - 3 | 559 | 5167173 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000288.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03005918.hg.1 | TC03000288.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005918.hg.1 | ENST00000332780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005918.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005918.hg.1 | ENST00000538729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005918.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000332780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000459846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000471811.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | ENST00000476495.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | NM_173546.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300152322.hg.1 | TC0300007367.hg.1 | Clariom D TC |
