Assay Details
Gene Symbol: PPM1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177983.2 | NP_817092.1 | 4 - 5 | 672 | 113 | |
GenBank mRNA | AK127593.1 | - | 4 - 5 | 538 | 113 | |
AK293141.1 | - | 5 - 6 | 466 | 113 | ||
AK312455.1 | - | 4 - 5 | 662 | 113 | ||
BC000057.2 | - | 4 - 5 | 521 | 3505271 | 113 | |
BC022061.2 | - | 4 - 5 | 518 | 2988852 | 113 | |
BT007454.1 | - | 4 - 5 | 411 | 113 | ||
CR457059.1 | - | 4 - 5 | 411 | 113 | ||
Y13936.1 | - | 4 - 5 | 435 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001685.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02027231.hg.1 | TC02001685.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027231.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027231.hg.1 | ENST00000350803.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027231.hg.1 | ENST00000544412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027231.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | TC02001685.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | ENST00000350803.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | ENST00000395543.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | ENST00000544412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027232.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | TC02001685.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | ENST00000350803.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | ENST00000395543.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | ENST00000544412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027230.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | TC02001685.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | ENST00000350803.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | ENST00000395543.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | ENST00000544412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027229.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188186.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188186.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188186.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188186.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188186.hg.1 | TC0200012076.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | ENST00000484925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012076.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188185.hg.1 | TC0200012076.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | ENST00000344034.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | ENST00000472077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | NM_177983.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188187.hg.1 | TC0200012076.hg.1 | Clariom D TC |
