Assay Details
| Gene Symbol: NCOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003743.4 | NP_003734.3 | 20 - 21 | 4411 | 79 | |
| XM_005264625.1 | XP_005264682.1 | 22 - 23 | 4687 | 79 | ||
| GenBank mRNA | AJ000881.1 | - | 20 - 21 | 4354 | 79 | |
| BC111533.1 | - | 19 - 20 | 4248 | 40027438 | 79 | |
| U40396.1 | - | 10 - 11 | 3069 | 79 | ||
| U90661.1 | - | 18 - 19 | 4195 | 79 | ||
More Information
Set Membership: |
|

NCOA1
Genomic Map