Assay Details
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004522.2 | NP_004513.1 | 12 - 13 | 1689 | 116 | |
NR_111932.1 | 5 - 6 | 711 | 116 | |||
XM_011511157.2 | XP_011509459.1 | 5 - 6 | 770 | 116 | ||
XM_017004062.1 | XP_016859551.1 | 12 - 13 | 1680 | 116 | ||
GenBank mRNA | AB011103.1 | - | 12 - 13 | 1662 | 116 | |
AF010146.1 | - | 2 - 3 | 233 | 116 | ||
BC110287.1 | - | 5 - 6 | 641 | 5165262 | 116 | |
BC127772.1 | - | 9 - 10 | 1013 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000927.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | TC02000927.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | ENST00000397413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | ENST00000414838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | ENST00000435030.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | ENST00000464066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013549.hg.1 | NM_004522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167251.hg.1 | TC0200009616.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
