Assay Details
| Gene Symbol: KIF5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004522.2 | NP_004513.1 | 4 - 5 | 791 | 149 | |
| XM_017004062.1 | XP_016859551.1 | 4 - 5 | 782 | 149 | ||
| GenBank mRNA | AB011103.1 | - | 4 - 5 | 764 | 149 | |
| BC002721.2 | - | 4 - 5 | 773 | 149 | ||
| BC017298.2 | - | 4 - 5 | 746 | 149 | ||
| BC025961.1 | - | 4 - 5 | 773 | 149 | ||
| BC127772.1 | - | 1 - 2 | 115 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013536.hg.1 | TC02000927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013536.hg.1 | ENST00000414838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013536.hg.1 | ENST00000435030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013536.hg.1 | NM_004522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |

KIF5C
Genomic Map