Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000237.hg.1 |
TC22001452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000237.hg.1 |
ENST00000316027.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000237.hg.1 |
ENST00000330423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000237.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000237.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
TC2200009187.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167089.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
TC2200009187.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167090.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000316027.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000330423.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000416740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000426208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
ENST00000608634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_001193414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009187.hg.1 |
NM_018943.2 |
Refseq |