Assay Details
| Gene Symbol: NUP62CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017681.2 | NP_060151.2 | 5 - 6 | 600 | 75 | |
| GenBank mRNA | AK000137.1 | - | 5 - 6 | 474 | 75 | |
| BC016327.1 | - | 5 - 6 | 597 | 4102213 | 75 | |
| KU178655.1 | - | 3 - 4 | 341 | 75 | ||
| KU178656.1 | - | 3 - 4 | 341 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001260.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | TC0X001260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | ENST00000372461.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | ENST00000372465.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | ENST00000372466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017029.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171295.hg.1 | TC0X00010465.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000372466.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000421752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000432145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | ENST00000484614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NM_017681.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010465.hg.1 | NR_033676.1 | Refseq |

NUP62CL
Genomic Map