Assay Details
Gene Symbol: KDM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014663.2 | NP_055478.2 | 11 - 12 | 1900 | 95 | |
XM_005271354.3 | XP_005271411.1 | 11 - 12 | 1937 | 95 | ||
XM_005271355.3 | XP_005271412.1 | 11 - 12 | 1855 | 95 | ||
XM_005271356.3 | XP_005271413.1 | 12 - 13 | 1976 | 95 | ||
XM_011542467.2 | XP_011540769.2 | 2 - 3 | 1008 | 95 | ||
XM_017002912.1 | XP_016858401.1 | 12 - 13 | 2045 | 95 | ||
GenBank mRNA | AB014577.1 | - | 11 - 12 | 1859 | 95 | |
BC002558.2 | - | 11 - 12 | 1868 | 3138875 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000549.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
negative regulation of autophagy
negative regulation of gene expression
cardiac muscle hypertrophy in response to stress
viral process
histone demethylation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
negative regulation of histone H3-K9 trimethylation
protein binding
zinc ion binding
ubiquitin protein ligase binding
histone demethylase activity
methylated histone binding
histone demethylase activity (H3-K36 specific)
transcription, DNA-templated
negative regulation of autophagy
negative regulation of gene expression
cardiac muscle hypertrophy in response to stress
viral process
histone demethylation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
negative regulation of histone H3-K9 trimethylation
protein binding
zinc ion binding
ubiquitin protein ligase binding
histone demethylase activity
methylated histone binding
histone demethylase activity (H3-K36 specific)
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01008881.hg.1 | TC01000549.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01008881.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01008881.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100157147.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100157147.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100157147.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100157147.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100157147.hg.1 | TC0100008073.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
