Assay Details
| Gene Symbol: CAPN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005632.2 | NP_005623.1 | 3 - 4 | 340 | 98 | |
| XM_011522630.2 | XP_011520932.1 | 3 - 4 | 401 | 98 | ||
| GenBank mRNA | U85647.1 | - | 3 - 4 | 340 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000015.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_005632.2,U85647.1,XM_011522630.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16000375.hg.1 | ENST00000219611.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000375.hg.1 | ENST00000397687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000375.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16000375.hg.1 | TC16000015.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008958.hg.1 | ENST00000565879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008958.hg.1 | ENST00000565879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144512.hg.1 | TC1600006469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600144513.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144513.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144513.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144513.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144513.hg.1 | TC1600006469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008958.hg.1 | ENST00000565879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008958.hg.1 | ENST00000565879.1 | Ensembl |

CAPN15
Genomic Map