Assay Details
| Gene Symbol: SAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000541.4 | NP_000532.2 | 12 - 13 | 1409 | 78 | |
| XM_011511593.2 | XP_011509895.1 | 8 - 9 | 1113 | 78 | ||
| XM_011511594.1 | XP_011509896.1 | 8 - 9 | 722 | 78 | ||
| XM_011511596.1 | XP_011509898.1 | 9 - 10 | 1316 | 78 | ||
| XM_017004641.1 | XP_016860130.1 | 11 - 12 | 1098 | 78 | ||
| XM_017004642.1 | XP_016860131.1 | 11 - 12 | 1098 | 78 | ||
| GenBank mRNA | AK302211.1 | - | 9 - 10 | 1316 | 78 | |
| BQ636894.1 | - | 4 - 5 | 323 | 78 | ||
| BU734369.1 | - | 4 - 5 | 78 | |||
| BX537502.1 | - | 5 - 6 | 1038 | 78 | ||
| BX641043.1 | - | 10 - 11 | 2240 | 78 | ||
| BX647821.1 | - | 12 - 13 | 3053 | 78 | ||
| BX647827.1 | - | 9 - 10 | 6207 | 78 | ||
| X12453.1 | - | 12 - 13 | 1242 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SAG
Genomic Map