Assay Details
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017018514.1 | XP_016874003.1 | 1 - 2 | 488 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017018514.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100016462.hg.1 | ENST00000420029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015996.hg.1 | ENST00000412556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012213.hg.1 | ENST00000445285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007542.hg.1 | ENST00000553684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000312175.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000445560.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000524663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000527348.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000528648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000530204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | ENST00000533166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_001143985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008049.hg.1 | NM_003860.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007542.hg.1 | ENST00000553684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015996.hg.1 | ENST00000412556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012213.hg.1 | ENST00000445285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016462.hg.1 | ENST00000420029.1 | Ensembl |
